Få mere at vide
4,4'-Dimethoxy Diphenyl Sulfide, TRC
Artikelnummer. 30377184
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377184 50 mg
30374434 500 mg
30368858 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30377184 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46099850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3393-77-9
Molekylær formel C14H14O2S
Molekylvægt (g/mol) 246.32
SMIL COc1ccc(Sc2ccc(OC)cc2)cc1
Tekniske data
Kemisk navn eller materiale 4,4'-Dimethoxy Diphenyl Sulfide
Mængde 50 mg
InChI formel InChI=1S/C14H14O2S/c1-15-11-3-7-13(8-4-11)17-14-9-5-12(16-2)6-10-14/h3-10H,1-2H3
Formel vægt 246.0715