missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-110625
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
345-92-6
C13H8F2O
218.20
MFCD00000353
LSQARZALBDFYQZ-UHFFFAOYSA-N
4,4'-difluorobenzophenone, bis 4-fluorophenyl methanone, bis 4-fluorophenyl-methanone, methanone, bis 4-fluorophenyl, p,p'-difluorobenzophenone, di-p-fluorophenyl ketone, bis p-fluorophenyl ketone, bis 4-fluorophenyl ketone, benzophenone, 4,4'-difluoro, unii-88bnc11b9c
9582
bis(4-fluorophenyl)methanone
FC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
This item is not returnable. View return policy
345-92-6
C13H8F2O
218.20
MFCD00000353
LSQARZALBDFYQZ-UHFFFAOYSA-N
4,4'-difluorobenzophenone, bis 4-fluorophenyl methanone, bis 4-fluorophenyl-methanone, methanone, bis 4-fluorophenyl, p,p'-difluorobenzophenone, di-p-fluorophenyl ketone, bis p-fluorophenyl ketone, bis 4-fluorophenyl ketone, benzophenone, 4,4'-difluoro, unii-88bnc11b9c
9582
bis(4-fluorophenyl)methanone
FC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 106°C to 109°C
Color White to Yellow
Boiling Point 200°C
Loss on Drying 0.25% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (FC6H4)2CO
Quantity 5 g
Formula Weight 218.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Difluorobenzophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Difluorobenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'