Få mere at vide
4,4-Difluoro-1,2,3,4-tetrahydroisoquinoline-1,3-dione, TRC
Artikelnummer. 30340898
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340898 100 mg
30334175 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30340898 Leverandør Toronto Research Chemicals Leverandørnr. TRCB426860100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,4-Difluoro-1,2,3,4-tetrahydroisoquinoline-1,3-dione
CAS 1443981-83-6
Molekylær formel C9H5NO2F2
Mængde 100 mg
InChI formel InChI=1S/C9H5F2NO2/c10-9(11)6-4-2-1-3-5(6)7(13)12-8(9)14/h1-4H,(H,12,13,14)
SMIL FC1(F)C(=O)NC(=O)c2ccccc12
IUPAC navn 4,4-difluoroisoquinoline-1,3-dione
Molekylvægt (g/mol) 197.14
Formel vægt 197.0288