Få mere at vide
4,4'-Dichlorodiphenyltrichloroethane - d8, TRC
Artikelnummer. 30135901
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135901 1 mg
30135087 5 mg
17948289 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30135901 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4341971MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93952-18-2
Molekylær formel C14 2H8 H Cl5
Molekylvægt (g/mol) 362.54
Synonym 3,3'-(2,2,2-Trichloroethylidene)bis[6-chlorobenzene-1,2,4,5-d4]
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-[2,2,2-trichloro-1-(4-chloro-2,3,5,6-tetradeuteriophenyl)ethyl]benzene
SMIL [2H]c1c([2H])c(C(c2c([2H])c([2H])c(Cl)c([2H])c2[2H])C(Cl)(Cl)Cl)c([2H])c([2H])c1Cl
Tekniske data
Kemisk navn eller materiale 4,4'-DDT D8
Mængde 1 mg
InChI formel InChI=1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H/i1D,2D,3D,4D,5D,6D,7D,8D
Formel vægt 359.9649