Få mere at vide
4,4'-Dichlorodiphenyldichloroethane - d8, TRC
Artikelnummer. 30373872
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30373872 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373872 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4341875MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,4'-DDD ring-d8
CAS 93952-20-6
Molekylær formel C14 2H8 H2 Cl4
Mængde 5 mg
Synonym 4,4'-DDD D8 (diphenyl D8), Benzene-1,2,4,5-d4, 3,3'-(2,2-dichloroethylidene)bis[6-chloro- (9CI), 4,4'-DDD D8, Benzene-1,2,4,5-d4, 3,3'-(2,2-dichloroethylidene)bis[6-chloro- (9CI)
InChI formel InChI=1S/C14H10Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13-14H/i1D,2D,3D,4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c(C(C(Cl)Cl)c2c([2H])c([2H])c(Cl)c([2H])c2[2H])c([2H])c([2H])c1Cl
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-[2,2-dichloro-1-(4-chloro-2,3,5,6-tetradeuteriophenyl)ethyl]benzene
Molekylvægt (g/mol) 328.09
Formel vægt 326.0039