Få mere at vide
4,4'-Dichlorobiphenyl-d8, CDN
Artikelnummer. 30316344
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30316344 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316344 Leverandør CDN Leverandørnr. CDND69180.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale PCB No. 15 D8
CAS 1219805-77-2
Molekylær formel C12 D8 Cl2
Mængde 0.01 g
Synonym 4,4'-Dichlorobiphenyl-d8, 4,4'-Dichlorobiphenyl-2,2',3,3',5,5',6,6'-d8, 4,4'-DiCB-d8, PCB No. 15 D8, PCB 15-d8
InChI formel InChI=1 S/C12H8Cl2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cl)c2c([2 H])c([2 H])c(Cl)c([2 H])c2[2 H]
IUPAC navn 1-chloro-4-(4-chloro-2,3,5,6-tetradeuteriophenyl)-2,3,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 231.1472
Formel vægt 230.0505 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity