missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dibromobiphenyl, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
4,4'-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
4,4′-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 164°C to 169°C |
Boiling Point | 355°C to 360°C |
Quantity | 50 g |
UN Number | UN3152 |
Beilstein | 2044701 |
Solubility Information | Insoluble in water. |
Formula Weight | 312.02 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,4'-Dibromobiphenyl |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'