Få mere at vide
4,4’-Di-N-acetylamino-diphenylsulfone-d8, TRC
Artikelnummer. 30363353
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30363353 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363353 Leverandør Toronto Research Chemicals Leverandørnr. TRCD30857750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,4'-Di-N-acetylamino-diphenylsulfone-d8
CAS 557794-37-3
Molekylær formel C16 2H8 H8 N2 O4 S
Mængde 50 mg
InChI formel InChI=1S/C16H16N2O4S/c1-11(19)17-13-3-7-15(8-4-13)23(21,22)16-9-5-14(6-10-16)18-12(2)20/h3-10H,1-2H3,(H,17,19)(H,18,20)/i3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)c2c([2H])c([2H])c(NC(=O)C)c([2H])c2[2H]
IUPAC navn N-[4-(4-acetamido-2,3,5,6-tetradeuteriophenyl)sulfonyl-2,3,5,6-tetradeuteriophenyl]acetamide
Molekylvægt (g/mol) 340.42
Formel vægt 340.1333