Få mere at vide
4-(4-Chlorophenyl)phenylacetic acid, TRC
Artikelnummer. 30366611
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366611 100 mg
30367034 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366611 Leverandør Toronto Research Chemicals Leverandørnr. TRCC428210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5525-72-4
Molekylær formel C14H11ClO2
Molekylvægt (g/mol) 246.69
IUPAC navn 2-[4-(4-chlorophenyl)phenyl]acetic acid
SMIL OC(=O)Cc1ccc(cc1)c2ccc(Cl)cc2
Tekniske data
Kemisk navn eller materiale 4-(4-Chlorophenyl)phenylacetic acid
Mængde 100 mg
InChI formel InChI=1S/C14H11ClO2/c15-13-7-5-12(6-8-13)11-3-1-10(2-4-11)9-14(16)17/h1-8H,9H2,(H,16,17)
Formel vægt 246.0448