Få mere at vide
4-(4-Chlorophenyl)cyclohexanol-d5, TRC
Artikelnummer. 30364021
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30364021 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364021 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37775210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Chlorophenyl)cyclohexanol-d5
CAS 1189961-66-7
Molekylær formel C12H10D5ClO
Mængde 10 mg
InChI formel InChI=1S/C12H15ClO/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-2,5-6,10,12,14H,3-4,7-8H2/i7D2,8D2,12D
SMIL [2H]C1([2H])CC(CC([2H])([2H])C1([2H])O)c2ccc(Cl)cc2
IUPAC navn 4-(4-chlorophenyl)-1,2,2,6,6-pentadeuteriocyclohexan-1-ol
Molekylvægt (g/mol) 215.73
Formel vægt 215.1125