Learn More
4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific™
Product Code. 11306945
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11306945 25 g Amber Glass Bottle 25g
10049491 1 g Amber Glass Bottle 1g
10497052 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11306945 Supplier Thermo Scientific Maybridge Supplier No. SB02037EB
 Restricted Item
Forventet forsendelse: 27-11-2026
Add to basket
Chemical Identifiers
CAS 95233-37-7
Molecular Formula C13H15ClO2
Molecular Weight (g/mol) 238.711
MDL Number MFCD06797626
InChI Key NXXDIEYTMQYWJU-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
PubChem CID 2735792
IUPAC Name 4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O
Specifications
CAS Min % 97.0
Quantity 25 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula ClC6H4C6H10CO2H
Formula Weight 238.71
Percent Purity 97%
Chemical Name or Material (E)-4-(4-Chlorophenyl)cyclohexanecarboxylic acid