Få mere at vide
4-(4-Chlorophenyl)-1-imidazol-1-yl-(butan-d5)-2-ol, TRC
Artikelnummer. 30361710
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30361710 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361710 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3795422.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Chlorophenyl)-1-imidazol-1-yl-(butan-d5)-2-ol
CAS 1185063-39-1
Molekylær formel C13 2H5 H10 Cl N2 O
Mængde 2.5 mg
Synonym 4-(4-Chlorophenyl)-1,1,2,3,3-pentadeuterio-1-imidazol-1-ylbutan-2-ol, 4-(4-Chlorophenyl)-1-imidazol-1-yl-(butan-d5)-2-ol
InChI formel InChI=1S/C13H15ClN2O/c14-12-4-1-11(2-5-12)3-6-13(17)9-16-8-7-15-10-16/h1-2,4-5,7-8,10,13,17H,3,6,9H2/i3D2,6D2,13D
SMIL [2H]C(O)(Cn1ccnc1)C([2H])([2H])C([2H])([2H])c2ccc(Cl)cc2
IUPAC navn 4-(4-chlorophenyl)-2,3,3,4,4-pentadeuterio-1-imidazol-1-ylbutan-2-ol
Molekylvægt (g/mol) 255.75
Formel vægt 255.1187