missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Chlorobenzoyl)piperidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Melting Point | 60°C to 63°C |
Color | Yellow |
Quantity | 1 g |
Beilstein | 158791 |
Solubility Information | Slightly soluble in water. |
IUPAC Name | 4-(4-chlorobenzoyl)piperidine |
Formula Weight | 223.7 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 4-(4-Chlorobenzoyl)piperidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'