missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzoyl)piperidine, 97%, Thermo Scientific™

Catalog Number p-7056006
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
53220-41-0
C12H14ClNO
223.70
MFCD04113538
IYGWDOXHCPQXKN-UHFFFAOYSA-N
4-4-chlorobenzoyl piperidine, 4-chlorophenyl piperidin-4-yl methanone, 4-4-chlorophenyl carbonyl piperidine, 4-chlorophenyl-4-piperidinyl methanone, 4-chlorophenyl-piperidin-4-yl-methanone, maybridge1_003174, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, 4-4-chlorobenzoyl-piperidine, 4-chlorophenyl 4-piperidyl ketone, 4-chlorophenyl piperidin-4-ylmethanone
2777115
ClC1=CC=C(C=C1)C(=O)C1CCNCC1
This item is not returnable. View return policy
53220-41-0
C12H14ClNO
223.70
MFCD04113538
IYGWDOXHCPQXKN-UHFFFAOYSA-N
4-4-chlorobenzoyl piperidine, 4-chlorophenyl piperidin-4-yl methanone, 4-4-chlorophenyl carbonyl piperidine, 4-chlorophenyl-4-piperidinyl methanone, 4-chlorophenyl-piperidin-4-yl-methanone, maybridge1_003174, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, 4-4-chlorobenzoyl-piperidine, 4-chlorophenyl 4-piperidyl ketone, 4-chlorophenyl piperidin-4-ylmethanone
2777115
ClC1=CC=C(C=C1)C(=O)C1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 60°C to 63°C
Color Yellow
Quantity 250 mg
Beilstein 158791
Solubility Information Slightly soluble in water.
IUPAC Name (4-chlorophenyl)-piperidin-4-ylmethanone
Formula Weight 223.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Chlorobenzoyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'