Få mere at vide
4-(4-Chloro-2-methylphenoxy-d3)butyric Acid, TRC
Artikelnummer. 30483444
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30483444 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483444 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1997290.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Chloro-2-methylphenoxy-d3)butyric Acid
CAS 1219803-42-5
Molekylær formel C11 D3 H10 Cl O3
Mængde 0.5 mg
Synonym 4-(4-Chloro-2-methylphenoxy-d3)butyric Acid, MCPB D3 (ring D3), 4-(4-Chloro-2-methylphenoxy)butanoic Acid (ring D3), 4-(4-Chloro-2-methylphenoxy-3,5,6-d3)butanoic acid
InChI formel InChI=1S/C11H13ClO3/c1-8-7-9(12)4-5-10(8)15-6-2-3-11(13)14/h4-5,7H,2-3,6H2,1H3,(H,13,14)/i4D,5D,7D
SMIL [2H]c1c([2H])c(OCCCC(=O)O)c(C)c([2H])c1Cl
IUPAC navn 4-(4-chloro-2,3,5-trideuterio-6-methylphenoxy)butanoic acid
Molekylvægt (g/mol) 231.6906
Formel vægt 231.0742