missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
4-(4-Chloro-2-methylphenoxy-d3)butyric Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(4-Chloro-2-methylphenoxy-d3)butyric Acid |
| CAS | 1219803-42-5 |
| Molekylær formel | C11 D3 H10 Cl O3 |
| Mængde | 5 mg |
| Synonym | 4-(4-Chloro-2-methylphenoxy-d3)butyric Acid, MCPB D3 (ring D3), 4-(4-Chloro-2-methylphenoxy)butanoic Acid (ring D3), 4-(4-Chloro-2-methylphenoxy-3,5,6-d3)butanoic acid |
| InChI formel | InChI=1S/C11H13ClO3/c1-8-7-9(12)4-5-10(8)15-6-2-3-11(13)14/h4-5,7H,2-3,6H2,1H3,(H,13,14)/i4D,5D,7D |
| SMIL | [2H]c1c([2H])c(OCCCC(=O)O)c(C)c([2H])c1Cl |
| IUPAC navn | 4-(4-chloro-2,3,5-trideuterio-6-methylphenoxy)butanoic acid |
| Molekylvægt (g/mol) | 231.6906 |
| Formel vægt | 231.0742 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?