missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Carbonyldibenzonitrile (4,4'-Dicyanobenzophenone), Mikromol™

Product Code. 30051010
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30051010 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051010 Supplier Mikromol Supplier No. MM1143.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4,4'-Carbonyldibenzonitrile (4,4'-Dicyanobenzophenone)
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4,4'-Carbonyldibenzonitrile
CAS 32446-66-5
Analyt- eller komponentnavne 4,4'-Carbonyldibenzonitrile
Forsendelsestilstand Room Temperature
Molekylær formel C15 H8 N2 O
Certificeringer/compliance ISO 9001
Synonym Benzonitrile, 4,4'-carbonylbis-, Benzonitrile, 4,4'-carbonyldi- (8CI), 4,4'-Carbonylbis[benzonitrile], 4,4'-Dicyanobenzophenone, 4-[(4-Cyanophenyl)carbonyl]benzonitrile, Bis(4-cyanophenyl) ketone, Bis(p-cyanophenyl) ketone, p,p'-Dicyanobenzophenone
InChI formel InChI=1S/C15H8N2O/c16-9-11-1-5-13(6-2-11)15(18)14-7-3-12(10-17)4-8-14/h1-8H
SMIL O=C(c1ccc(cc1)C#N)c2ccc(cc2)C#N
IUPAC navn 4-(4-cyanobenzoyl)benzonitrile
Molekylvægt (g/mol) 232.24
Mængde 25 mg
Formel vægt 232.0637 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.