missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Biphenylyl)-2-cyclopropylthiazole, 97%, Thermo Scientific™

Catalog Number 15424518
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1421261-24-6
C18H15NS
277.39
MFCD22682945
ZGFVNJMOZOGCDK-UHFFFAOYSA-N
4-4-biphenylyl-2-cyclopropylthiazole, 4-1,1'-biphenyl-4-yl-2-cyclopropyl-1,3-thiazole
73996310
C1CC1C1=NC(=CS1)C1=CC=C(C=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1421261-24-6
C18H15NS
277.39
MFCD22682945
ZGFVNJMOZOGCDK-UHFFFAOYSA-N
4-4-biphenylyl-2-cyclopropylthiazole, 4-1,1'-biphenyl-4-yl-2-cyclopropyl-1,3-thiazole
73996310
C1CC1C1=NC(=CS1)C1=CC=C(C=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
IUPAC Name 4-{[1,1'-biphenyl]-4-yl}-2-cyclopropyl-1,3-thiazole
Formula Weight 353.48
Percent Purity 97%
Chemical Name or Material 4-(4-Biphenylyl)-2-cyclopropylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'