Få mere at vide
4-(4-Aminophenyl)morpholin-3-one, Mikromol™
Artikelnummer. 30051194
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051194 100 mg
30049437 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051194 Leverandør Mikromol Leverandørnr. MM3629.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(4-Aminophenyl)morpholin-3-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Aminophenyl)-3-morpholinone
CAS 438056-69-0
Analyt- eller komponentnavne 4-(4-Aminophenyl)-3-morpholinone
Forsendelsestilstand Room Temperature
Molekylær formel C10 H12 N2 O2
Certificeringer/compliance ISO 17025
Synonym 4-(3-Oxo-4-morpholinyl)aniline, 4-(4-Aminophenyl)-3-morpholinone, 4-(4-Aminophenyl)morpholin-3-one
InChI formel InChI=1S/C10H12N2O2/c11-8-1-3-9(4-2-8)12-5-6-14-7-10(12)13/h1-4H,5-7,11H2
SMIL Nc1ccc(cc1)N2CCOCC2=O
IUPAC navn 4-(4-aminophenyl)morpholin-3-one
Molekylvægt (g/mol) 192.21
Mængde 100 mg
Formel vægt 192.0899 g/mol