Få mere at vide
4-(4-Aminophenyl)-3-morpholinone-d4, TRC
Artikelnummer. 30316961
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30316961 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316961 Leverandør Toronto Research Chemicals Leverandørnr. TRCA625992100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Aminophenyl)-3-morpholinone-d4
CAS 1329837-80-0
Molekylær formel C10H8D4N2O2
Mængde 100 mg
Synonym 4-(4-Aminophenyl)morpholin-3-one, 4-(3-Oxo-4-morpholinyl)aniline-d4
InChI formel InChI=1S/C10H12N2O2/c11-8-1-3-9(4-2-8)12-5-6-14-7-10(12)13/h1-4H,5-7,11H2/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(N2CCOCC2=O)c([2H])c([2H])c1N
IUPAC navn 4-(4-amino-2,3,5,6-tetradeuteriophenyl)morpholin-3-one
Molekylvægt (g/mol) 196.24
Formel vægt 196.115