Få mere at vide
[4-(4-Aminophenoxy)(2-pyridyl)]-N-(methyl-d3)carboxamide, TRC
Artikelnummer. 30325402
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30325402 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325402 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61894210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4-(4-Aminophenoxy)(2-pyridyl))-N-(methyl-d3)carboxamide
CAS 1189975-18-5
Molekylær formel C13H10D3N3O2
Mængde 10 mg
Synonym 2-Pyridinecarboxamide, 4-(4-aminophenoxy)-N-(methyl-d3)-, 4-(4-Aminophenoxy)-N-(methyl-d3)-2-pyridinecarboxamide, [4-(4-Aminophenoxy)(2-pyridyl)]-N-(methyl-d3)carboxamide, 4-(4-Aminophenoxy)-N-methylpyridine-2-carboxamide-D3 (N-methyl-D3), 4-(4-Aminophenoxy)-N-methylpyridine-2-carboxamide-d3, 4-(4-Aminophenoxy)pyridine-2-carboxylic acid trideuteriomethylamide, 4-[[2-(Methyl-d3-carbamoyl)pyridin-4-yl]oxy]aniline
InChI formel InChI=1S/C13H13N3O2/c1-15-13(17)12-8-11(6-7-16-12)18-10-4-2-9(14)3-5-10/h2-8H,14H2,1H3,(H,15,17)/i1D3
SMIL [2H]C([2H])([2H])NC(=O)c1cc(Oc2ccc(N)cc2)ccn1
IUPAC navn 4-(4-aminophenoxy)-N-(trideuteriomethyl)pyridine-2-carboxamide
Molekylvægt (g/mol) 246.28
Formel vægt 246.1196