Få mere at vide
4,4'-[9H-Carbazole-3,6-diyldi-(1E)-2,1-ethenediyl]bis[1-methyl-pyridinium Diiodide (>90%), TRC
Artikelnummer. 30353405
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353405 50 mg
30360987 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30353405 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17607050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 647859-37-8
Molekylær formel C28 H25 N3 . 2 I
Molekylvægt (g/mol) 657.327
Synonym 4,4'-[9H-Carbazole-3,6-diyldi-(1E)-2,1-ethenediyl]bis[1-methyl-pyridinium Iodide (1:2), 3,6-Bis(1-methyl-4-vinylpyridinium iodine) Carbazole
IUPAC navn 3,6-bis[2-(1-methylpyridin-1-ium-4-yl)ethenyl]-9H-carbazole;diiodide
SMIL [I-].[I-].C[n+]1ccc(C=Cc2ccc3[nH]c4ccc(C=Cc5cc[n+](C)cc5)cc4c3c2)cc1
Tekniske data
Kemisk navn eller materiale 4,4'-[9H-Carbazole-3,6-diyldi-(1E)-2,1-ethenediyl]bis-1-methyl-pyridinium Diiodide
Mængde 50 mg
InChI formel InChI=1S/C28H24N3.2HI/c1-30-15-11-21(12-16-30)3-5-23-7-9-27-25(19-23)26-20-24(8-10-28(26)29-27)6-4-22-13-17-31(2)18-14-22;;/h3-20H,1-2H3;2*1H/q+1;;/p-1
Formel vægt 657.014