missing translation for 'onlineSavingsMsg'
Få mere at vide

4,4,5,5,5-Pentafluoropentanol, TRC

Artikelnummer. 30422597
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422597 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30422597 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2703252G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4,4,5,5,5-Pentafluoropentanol
CAS 148043-73-6
Molekylær formel C5H7F5O
Mængde 2 g
Synonym 4,4,5,5,5-Pentafluoro-1-pentanol
InChI formel InChI=1S/C5H7F5O/c6-4(7,2-1-3-11)5(8,9)10/h11H,1-3H2
SMIL OCCCC(F)(F)C(F)(F)F
IUPAC navn 4,4,5,5,5-pentafluoropentan-1-ol
Molekylvægt (g/mol) 178.1
Formel vægt 178.0417
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.