missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5,5-Pentafluoropentanol, 94%, Thermo Scientific Chemicals

Catalog Number p-7025823
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
148043-73-6
C5H7F5O
178.102
MFCD00153224
QROUUECTKRZFHF-UHFFFAOYSA-N
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
547967
4,4,5,5,5-pentafluoropentan-1-ol
C(CC(C(F)(F)F)(F)F)CO
This item is not returnable. View return policy
148043-73-6
C5H7F5O
178.102
MFCD00153224
QROUUECTKRZFHF-UHFFFAOYSA-N
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
547967
4,4,5,5,5-pentafluoropentan-1-ol
C(CC(C(F)(F)F)(F)F)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.35
Boiling Point 135°C
Flash Point 63°C (145°F)
Linear Formula CF3CF2(CH2)3OH
Refractive Index 1.334
Quantity 2 g
Formula Weight 178.1
Percent Purity 94%
Chemical Name or Material 4,4,5,5,5-Pentafluoropentanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'