missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,4,5,5,5-Pentafluoropentanol, 94%

Brand:  Alfa Aesar™ B21512.04

Additional Details : CAS Number : 148043-73-6 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11497064

  • 224.00 DKK / 2g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

148043-73-6
178.102
QROUUECTKRZFHF-UHFFFAOYSA-N
547967
C(CC(C(F)(F)F)(F)F)CO
C5H7F5O
MFCD00153224
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
4,4,5,5,5-pentafluoropentan-1-ol

Specifications

4,4,5,5,5-Pentafluoropentanol
63°C (145°F)
135°C
148043-73-6
CF3CF2(CH2)3OH
4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
C(CC(C(F)(F)F)(F)F)CO
178.102
178.1
1.35
1.334
2g
C5H7F5O
MFCD00153224
QROUUECTKRZFHF-UHFFFAOYSA-N
4,4,5,5,5-pentafluoropentan-1-ol
547967
94%
Videos
SDS Documents