Få mere at vide
4,4,4-Trifluoro-2-(aminomethylphenyl)methylene-3-oxo-butanoic Acid Ethyl Ester, TRC
Artikelnummer. 30453876
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453876 50 mg
30449043 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30453876 Leverandør Toronto Research Chemicals Leverandørnr. TRCT78990050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,4,4-Trifluoro-2-(aminomethylphenyl)methylene-3-oxo-butanoic Acid Ethyl Ester
CAS 1798421-25-6
Molekylær formel C14H14F3NO3
Mængde 50 mg
Synonym 2-[Amino-(4-methylphenyl)]methylene-4,4,4-trifluoro-3-oxo-butanoic Acid Ethyl Ester
InChI formel InChI=1S/C14H14F3NO3/c1-3-21-13(20)10(12(19)14(15,16)17)11(18)9-6-4-8(2)5-7-9/h4-7H,3,18H2,1-2H3/b11-10-
SMIL CCOC(=O)\C(=C(/N)\c1ccc(C)cc1)\C(=O)C(F)(F)F
IUPAC navn ethyl (2Z)-2-[amino-(4-methylphenyl)methylidene]-4,4,4-trifluoro-3-oxobutanoate
Molekylvægt (g/mol) 301.26
Formel vægt 301.0926