Learn More
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione |
| CAS | 582-65-0 |
| Molecular Formula | C10H6F4O2 |
| Quantity | 5 g |
| Synonym | 1,3-Butanedione','4,4,4-trifluoro-1-(p-fluorophenyl)- (6CI','8CI), 4,4,4-Trifluoro-1-(4-fluorophenyl)-1,3-butanedione (ACI), 1,1,1-Trifluoro-4-(4-fluorophenyl)-2,4-butanedione, 1-(4-Fluorophenyl)-4,4,4-trifluoro-1,3-butanedione, 1-(4′-Fluorophenyl)-4,4,4-trifluoro-1,3-butanedione, 4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione |
| InChI Formula | InChI=1S/C10H6F4O2/c11-7-3-1-6(2-4-7)8(15)5-9(16)10(12,13)14/h1-4H,5H2 |
| SMILES | O=C(C1=CC=C(F)C=C1)CC(C(F)(F)F)=O |
| IUPAC Name | 4,4,4-trifluoro-1-(4-fluorophenyl)butane-1,3-dione |
| Molecular Weight (g/mol) | 234.15 |
| Formula Weight | 234.03 |