Få mere at vide
4,4',4' '-Tri-tert-butyl-2,2':6',2' '-terpyridine, TRC
Artikelnummer. 30428437
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428437 10 mg
30410353 100 mg
30422235 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428437 Leverandør Toronto Research Chemicals Leverandørnr. TRCT11509010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 115091-29-7
Molekylær formel C27H35N3
Molekylvægt (g/mol) 401.59
Synonym 4,4',4' '-Tris(1,1-dimethylethyl)-2,2':6',2' '-terpyridine, 2,6-Bis[4-(tert-butyl)pyridin-2-yl)-4-(tert-butyl)pyridine
IUPAC navn 4-tert-butyl-2,6-bis(4-tert-butylpyridin-2-yl)pyridine
SMIL CC(C)(C)c1ccnc(c1)c2cc(cc(n2)c3cc(ccn3)C(C)(C)C)C(C)(C)C
Tekniske data
Kemisk navn eller materiale 4,4',4' '-Tri-tert-butyl-2,2':6',2' '-terpyridine
Mængde 10 mg
InChI formel InChI=1S/C27H35N3/c1-25(2,3)18-10-12-28-21(14-18)23-16-20(27(7,8)9)17-24(30-23)22-15-19(11-13-29-22)26(4,5)6/h10-17H,1-9H3
Formel vægt 401.2831