Få mere at vide
4-[4-[4-(Methoxymethoxy)phenyl]-1-piperazinyl]benzenamine-d4, TRC
Artikelnummer. 30483545
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30483545 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483545 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2643425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[4-[4-(Methoxymethoxy)phenyl]-1-piperazinyl]benzenamine-d4
CAS 1246816-20-5
Molekylær formel C18H19D4N3O2
Mængde 5 mg
Synonym 1-(4-Aminophenyl)-4-(4-Methoxymethyl-4-hydroxyphenyl)piperazine-d4
InChI formel InChI=1S/C18H23N3O2/c1-22-14-23-18-8-6-17(7-9-18)21-12-10-20(11-13-21)16-4-2-15(19)3-5-16/h2-9H,10-14,19H2,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(N2CCN(CC2)c3ccc(OCOC)cc3)c([2H])c([2H])c1N
IUPAC navn 2,3,5,6-tetradeuterio-4-[4-[4-(methoxymethoxy)phenyl]piperazin-1-yl]aniline
Molekylvægt (g/mol) 317.42
Formel vægt 317.2041