Få mere at vide
4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-Alpha,Alpha-dimethyl-benzeneacetic Acid, TRC
Artikelnummer. 30371701
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371701 10 mg
30369160 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30371701 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49202010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1187954-57-9
Molekylær formel C32 H37 N O3
Molekylvægt (g/mol) 483.64
Synonym Benzeneacetic acid, 4-[4-[4-(diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-α,α-dimethyl-, 2-[4-[(1RS)-4-[4-(Diphenylmethyl-idene)piperidin-1-yl]-1-hydroxybutyl]phenyl]-2-methylpropanoic Acid
IUPAC navn 2-[4-[4-(4-benzhydrylidenepiperidin-1-yl)-1-hydroxybutyl]phenyl]-2-methylpropanoic acid
SMIL CC(C)(C(=O)O)c1ccc(cc1)C(O)CCCN2CCC(=C(c3ccccc3)c4ccccc4)CC2
Tekniske data
Kemisk navn eller materiale 4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-alpha,alpha-dimethyl-benzeneacetic acid
Mængde 10 mg
InChI formel InChI=1S/C32H37NO3/c1-32(2,31(35)36)28-17-15-24(16-18-28)29(34)14-9-21-33-22-19-27(20-23-33)30(25-10-5-3-6-11-25)26-12-7-4-8-13-26/h3-8,10-13,15-18,29,34H,9,14,19-23H2,1-2H3,(H,35,36)
Formel vægt 483.2773