missing translation for 'onlineSavingsMsg'
Learn More

4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine, 97%, Thermo Scientific™

Catalog Number p-3882137
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
364794-85-4
C15H24BNO3S
309.23
MFCD11841079
AMUMGAXTCHBNPU-UHFFFAOYSA-N
4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-ylmethyl morpholine, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 2-morpholin-4-ylmethyl thiophene-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-yl methyl morpholine, amtb834, 2-morpholin-4-ylmethyl thiophene-4-boronic acid pinacol ester, 2-morpholin-4-ylmethyl thiophene-4-boronic acid,pinacol ester
23438311
4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]morpholine
CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1
This item is not returnable. View return policy
364794-85-4
C15H24BNO3S
309.23
MFCD11841079
AMUMGAXTCHBNPU-UHFFFAOYSA-N
4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-ylmethyl morpholine, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl morpholine, 2-morpholin-4-ylmethyl thiophene-4-boronic acid, pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thien-2-yl methyl morpholine, amtb834, 2-morpholin-4-ylmethyl thiophene-4-boronic acid pinacol ester, 2-morpholin-4-ylmethyl thiophene-4-boronic acid,pinacol ester
23438311
4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]morpholine
CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 309.24
Percent Purity 97%
Chemical Name or Material 4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'