Få mere at vide
4,4'-(1,3,3-Trimethyl-1-propene-1,3-diyl)bisphenol, TRC
Artikelnummer. 30456224
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30456224 25 mg -
30454637 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456224 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79665525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57244-54-9
Molekylær formel C18H20O2
Molekylvægt (g/mol) 268.35
SMIL CC(=CC(C)(C)C1=CC=C(C=C1)O)C2=CC=C(C=C2)O
Tekniske data
Kemisk navn eller materiale 4,4'-(1,3,3-Trimethyl-1-propene-1,3-diyl)bisphenol
Mængde 25 mg
InChI formel InChI=1S/C18H20O2/c1-13(14-4-8-16(19)9-5-14)12-18(2,3)15-6-10-17(20)11-7-15/h4-12,19-20H,1-3H3
Formel vægt 268.1463