Få mere at vide
4-((3R,5S)-5-([1,1'-Biphenyl]-4-ylmethyl)-3-methyl-2-oxopyrrolidin-1-yl)-4-oxobutanoic Acid, TRC
Artikelnummer. 30134404
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134404 100 mg
30009541 10 mg
30005505 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134404 Leverandør Toronto Research Chemicals Leverandørnr. TRCB398720100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2216746-87-9
Molekylær formel C22 H23 N O4
Molekylvægt (g/mol) 365.422
Synonym Valsartan Impurity
IUPAC navn 4-[(3R,5S)-3-methyl-2-oxo-5-[(4-phenylphenyl)methyl]pyrrolidin-1-yl]-4-oxo-butanoic acid
SMIL C[C@@H]1C[C@@H](Cc2ccc(cc2)c3ccccc3)N(C(=O)CCC(=O)O)C1=O
Tekniske data
Kemisk navn eller materiale 4-[(3R,5S)-3-methyl-2-oxo-5-[(4-phenylphenyl)methyl]pyrrolidin-1-yl]-4-oxo-butanoic acid
Mængde 100 mg
InChI formel InChI=1S/C22H23NO4/c1-15-13-19(23(22(15)27)20(24)11-12-21(25)26)14-16-7-9-18(10-8-16)17-5-3-2-4-6-17/h2-10,15,19H,11-14H2,1H3,(H,25,26)/t15-,19+/m1/s1
Formel vægt 365.1627