missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7032123
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
874839-13-1
C12H7ClF3NO3S
337.697
MFCD07774164
FDAYBQVWHDJJOL-UHFFFAOYSA-N
4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
45594320
4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F
This item is not returnable. View return policy
874839-13-1
C12H7ClF3NO3S
337.697
MFCD07774164
FDAYBQVWHDJJOL-UHFFFAOYSA-N
4-3-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, chloro 4-3-trifluoromethyl 2-pyridyloxy phenyl sulfone, 4-3-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
45594320
4-[3-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
C1=CC(=C(N=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 63°C to 65°C
Quantity 250 mg
UN Number UN3261
Formula Weight 337.71
Percent Purity 97%
Chemical Name or Material 4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'