missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Pyridinyl)-2-pyrimidinamine, TRC

Product Code. 30412659
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30412659 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412659 Supplier Toronto Research Chemicals Supplier No. TRCP991700500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-(Pyridin-3-yl)pyrimidin-2-ylamine
CAS 66521-66-2
Molekylær formel C9 H8 N4
Mængde 500 mg
Synonym 2-Pyrimidinamine','4-(3-pyridinyl)-, 4-(3-Pyridinyl)-2-pyrimidinamine, 2-Amino-4-(3-pyridinyl)pyrimidine, 4-(3-Pyridyl)-2-pyrimidinamine, 4-(Pyridin-3-yl)pyrimidin-2-amine
InChI formel InChI=1S/C9H8N4/c10-9-12-5-3-8(13-9)7-2-1-4-11-6-7/h1-6H,(H2,10,12,13)
SMIL Nc1nccc(n1)c2cccnc2
IUPAC navn 4-pyridin-3-ylpyrimidin-2-amine
Molekylvægt (g/mol) 172.19
Formel vægt 172.0749
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.