missing translation for 'onlineSavingsMsg'
Få mere at vide

4-[(3-Methylphenyl)amino]pyridin-3-sulfonamid, Mikromol™

Artikelnummer. 17911016
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17911016 25 mg
17901976 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17911016 Supplier Mikromol Supplier No. MM1283.020025
 Begrænset vare
Forventet forsendelse: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol – din pålidelige kilde til farmaceutiske referencestandarder af høj kvalitet. Understøttelse af nøjagtig, overensstemmende analyser med ISO 17034-akkrediterede materialer, der er designet til at sikre tillid til hvert resultat.

4-[(3-Methylphenyl)amino]pyridin-3-sulfonamid
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide
CAS 72811-73-5
Analyt- eller komponentnavne 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide
Forsendelsestilstand Stuetemperatur
Molekylær formel C12 H13 N3 O2 S
Certificeringer/compliance ISO 17025
Synonym Torasemide Imp. B (EP), Torsemide USP Related Compound A, Torsemide USP RC A, 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide, Torasemide, Anhydrous Imp. B (EP), Torsemide Related Compound A, Torasemide, Anhydrous Impurity B, Torasemide Impurity B
InChI formel InChI=1S/C12H13N3O2S/c1-9-3-2-4-10(7-9)15-11-5-6-14-8-12(11)18(13,16)17/h2-8H,1H3,(H,14,15)(H2,13,16,17)
SMIL O=S(=O)(N)C=1C=NC=CC1NC2=CC=CC(=C2)C
IUPAC navn 4-(3-methylanilino)pyridin-3-sulfonamid
Molekylvægt (g/mol) 263.32
Mængde 25 mg
Formel vægt 263,0728 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.