Få mere at vide
4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide, Mikromol™
Artikelnummer. 17901976
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17901976 100 mg
17911016 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17901976 Leverandør Mikromol Leverandørnr. MM1283.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide
CAS 72811-73-5
Analyt- eller komponentnavne 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide
Forsendelsestilstand Room Temperature
Molekylær formel C12 H13 N3 O2 S
Certificeringer/compliance ISO 17025
Synonym Torasemide Imp. B (EP), Torsemide USP Related Compound A, Torsemide USP RC A, 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide, Torasemide, Anhydrous Imp. B (EP), Torsemide Related Compound A, Torasemide, Anhydrous Impurity B, Torasemide Impurity B
InChI formel InChI=1S/C12H13N3O2S/c1-9-3-2-4-10(7-9)15-11-5-6-14-8-12(11)18(13,16)17/h2-8H,1H3,(H,14,15)(H2,13,16,17)
SMIL O=S(=O)(N)C=1C=NC=CC1NC2=CC=CC(=C2)C
IUPAC navn 4-(3-methylanilino)pyridine-3-sulfonamide
Molekylvægt (g/mol) 263.32
Mængde 100 mg
Formel vægt 263.0728 g/mol