Få mere at vide
4-(3-Methoxypropoxy)-2,3-dimethylpyridin-1-ium-1-olate, TRC
Artikelnummer. 30470275
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470275 100 mg
30470075 500 mg
30478173 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30470275 Leverandør Toronto Research Chemicals Leverandørnr. TRCM221163100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 117977-18-1
Molekylær formel C11H17NO3
Molekylvægt (g/mol) 211.25
Synonym 4-(3-Methoxypropoxy)-2,3-dimethylpyridine N-Oxide
IUPAC navn 4-(3-methoxypropoxy)-2,3-dimethyl-1-oxidopyridin-1-ium
SMIL COCCCOc1cc[n+]([O-])c(C)c1C
Tekniske data
Kemisk navn eller materiale 4-(3-Methoxypropoxy)-2,3-dimethylpyridin-1-ium-1-olate
Mængde 100 mg
InChI formel InChI=1S/C11H17NO3/c1-9-10(2)12(13)6-5-11(9)15-8-4-7-14-3/h5-6H,4,7-8H2,1-3H3
Formel vægt 211.1208