missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Hydroxy-1-propynyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number 15459395
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1449133-12-3
C15H19BO3
258.124
MFCD16294528
SYRFTFKBNBRKRB-UHFFFAOYSA-N
3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-prop-2-yn-1-ol, 4-3-hydroxy-1-propynyl benzeneboronic acid pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2-propyn-1-ol, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-yn-1-ol, amtb208
71306516
3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-yn-1-ol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CCO
This item is not returnable. View return policy
1449133-12-3
C15H19BO3
258.124
MFCD16294528
SYRFTFKBNBRKRB-UHFFFAOYSA-N
3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-prop-2-yn-1-ol, 4-3-hydroxy-1-propynyl benzeneboronic acid pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2-propyn-1-ol, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-yn-1-ol, amtb208
71306516
3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-yn-1-ol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C#CCO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 258.12
Percent Purity 95%
Chemical Name or Material 4-(3-Hydroxy-1-propynyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'