missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Fluorophenyl)-3-thiosemicarbazide, 97%, Thermo Scientific™

Catalog Number 11382547
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
70619-48-6
C7H8FN3S
185.22
MFCD00041286
HLLFMTUWRLDCPF-UHFFFAOYSA-N
n-3-fluorophenyl hydrazinecarbothioamide, 4-3-fluorophenyl-3-thiosemicarbazide, 3-amino-1-3-fluorophenyl thiourea, 1-amino-3-3-fluorophenyl thiourea, 3-fluorophenyl amino hydrazinomethane-1-thione, 4-m-fluorophenyl thiosemicarbazide, 1-azanyl-3-3-fluorophenyl thiourea, n-3-fluorophenyl-1-hydrazinecarbothioamide
2737499
1-amino-3-(3-fluorophenyl)thiourea
C1=CC(=CC(=C1)F)NC(=S)NN
This item is not returnable. View return policy
70619-48-6
C7H8FN3S
185.22
MFCD00041286
HLLFMTUWRLDCPF-UHFFFAOYSA-N
n-3-fluorophenyl hydrazinecarbothioamide, 4-3-fluorophenyl-3-thiosemicarbazide, 3-amino-1-3-fluorophenyl thiourea, 1-amino-3-3-fluorophenyl thiourea, 3-fluorophenyl amino hydrazinomethane-1-thione, 4-m-fluorophenyl thiosemicarbazide, 1-azanyl-3-3-fluorophenyl thiourea, n-3-fluorophenyl-1-hydrazinecarbothioamide
2737499
1-amino-3-(3-fluorophenyl)thiourea
C1=CC(=CC(=C1)F)NC(=S)NN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∽160°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 2804712
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material 4-(3-Fluorophenyl)-3-thiosemicarbazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'