missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chloropropyl)morpholine, 95%, Thermo Scientific Chemicals

Catalog Number p-7032883
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7357-67-7
C7H14ClNO
163.645
MFCD00039714
PIAZYBLGBSMNLX-UHFFFAOYSA-N
4-3-chloropropyl morpholine, n-3-chloropropyl morpholine, morpholine, 4-3-chloropropyl, 3-morpholinopropyl chloride, 1-chloro-3-morpholinopropane, 4-3-chloropropyl-morpholine, 1-chloro-3-morpholin-4-yl propane, 4-3-chloro-propyl-morpholine, morpholine, 4-3-chloropropyl-, hydrochloride
95834
4-(3-chloropropyl)morpholine
C1COCCN1CCCCl
This item is not returnable. View return policy
7357-67-7
C7H14ClNO
163.645
MFCD00039714
PIAZYBLGBSMNLX-UHFFFAOYSA-N
4-3-chloropropyl morpholine, n-3-chloropropyl morpholine, morpholine, 4-3-chloropropyl, 3-morpholinopropyl chloride, 1-chloro-3-morpholinopropane, 4-3-chloropropyl-morpholine, 1-chloro-3-morpholin-4-yl propane, 4-3-chloro-propyl-morpholine, morpholine, 4-3-chloropropyl-, hydrochloride
95834
4-(3-chloropropyl)morpholine
C1COCCN1CCCCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 g
Formula Weight 163.65
Percent Purity 95%
Chemical Name or Material 4-(3-Chloropropyl)morpholine
compliance-icons
Product Identifier
  • 4-(3-Chloropropyl)morpholine
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'