Få mere at vide
4-(3-Chlorophenyl)-4-oxo-2-butenoic Acid, TRC
Artikelnummer. 30366526
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366526 1 g
30366173 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366526 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3751201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(3-Chlorophenyl)-4-oxo-2-butenoic Acid
CAS 191015-04-0
Molekylær formel C10H7ClO3
Mængde 1 g
InChI formel InChI=1S/C10H7ClO3/c11-8-3-1-2-7(6-8)9(12)4-5-10(13)14/h1-6H,(H,13,14)/b5-4+
SMIL OC(=O)\C=C\C(=O)c1cccc(Cl)c1
IUPAC navn (E)-4-(3-chlorophenyl)-4-oxobut-2-enoic acid
Molekylvægt (g/mol) 210.61
Formel vægt 210.0084