Få mere at vide
4-(3-Chlorophenyl)-1-[3-(3-oxo-2,3-dihydro-1,2,4-triazolo[4,3-a]pyridin-2-yl)propyl]piperazine N1-Oxide Hydrochloride (Trazodone N-Oxide Hydrochloride), Mikromol™
Beskrivelse
4-(3-Chlorophenyl)-1-[3-(3-oxo-2,3-dihydro-1,2,4-triazolo[4,3-a]pyridin-2-yl)propyl]piperazine N1-Oxide Hydrochloride (Trazodone N-Oxide Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(3-Chlorophenyl)-1-[3-(3-oxo-2,3-dihydro-1,2,4-triazolo[4,3-a]pyridin-2-yl)propyl]piperazine N1-Oxide Hydrochloride (Trazodone N-Oxide Hydrochloride) |
| CAS | 55290-66-9 |
| Analyt- eller komponentnavne | 4-(3-Chlorophenyl)-1-[3-(3-oxo-2,3-dihydro-1,2,4-triazolo[4,3-a]pyridin-2-yl)propyl]piperazine N1-Oxide Hydrochloride (Trazodone N-Oxide Hydrochloride) |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C19 H22 Cl N5 O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 4-(3-Chlorophenyl)-1-[3-(3-oxo-2,3-dihydro-1,2,4-triazolo[4,3-a]pyridin-2-yl)propyl]piperazine N1-Oxide Hydrochloride, Trazodone Hydrochloride Imp. A (BP) as Hydrochloride, Trazodone N-Oxide Hydrochloride, Trazodone Hydrochloride Impurity A, Trazodone Impurity A |
| InChI formel | InChI=1S/C19H22ClN5O2.ClH/c20-16-5-3-6-17(15-16)22-10-13-25(27,14-11-22)12-4-9-24-19(26)23-8-2-1-7-18(23)21-24;/h1-3,5-8,15H,4,9-14H2;1H |
| SMIL | Cl.[O-][N+]1(CCCN2N=C3C=CC=CN3C2=O)CCN(CC1)c4cccc(Cl)c4 |
| IUPAC navn | 2-[3-[4-(3-chlorophenyl)-1-oxidopiperazin-1-ium-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride |
| Vis mere |