missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-chloro-2-pyridinyl)morpholine, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 54231-36-6 |
CAS Min % | 97.0 |
Assay Percent Range | 97+% |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H11ClN2O |
MDL Number | MFCD00498941 |
Quantity | 5 g |
Synonym | 4-3-chloropyridin-2-yl morpholine, 3-chloro-2-morpholinopyridine, 3-chloro-2-4-morpholino pyridine, maybridge2_000531, ksc915s6h, 2-morpholino-3-chloropyridine, 4-3-chloro-2-pyridyl morpholine, 3-chloro-2-morpholin-4-ylpyridine, 4-3-chloro-2-pyridinyl morpholine, 4-3-chloropyridin-2-yl ;morpholine |
InChI Key | KCKQIVLNKGDTGJ-UHFFFAOYSA-N |
SMILES | C1COCCN1C2=C(C=CC=N2)Cl |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'