Få mere at vide
4-(3-Carboxy-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinolin-7-yl)-1-methylpiperazine 1-Oxide (Pefloxacin N-Oxide), Mikromol™
Artikelnummer. 30052131
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30052131 25 mg
1 options
Artikelnummer. 30052131 Leverandør Mikromol Leverandørnr. MM3495.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(3-Carboxy-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinolin-7-yl)-1-methylpiperazine 1-Oxide (Pefloxacin N-Oxide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pefloxacin N-Oxide
CAS 85145-21-7
Analyt- eller komponentnavne Pefloxacin N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C17 H20 F N3 O4
Certificeringer/compliance ISO 9001
Synonym 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-4-oxido-1-piperazinyl)-4-oxo-, 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, N-oxide, Pefloxacin N-oxide, Pefloxacin Mesilate Dihydrate Imp. D (EP), Pefloxacin Mesilate Imp. D (EP), Pefloxacin Mesilate Dihydrate Impurity D, Pefloxacin Impurity D
InChI formel InChI=1S/C17H20FN3O4/c1-3-19-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)19)20-4-6-21(2,25)7-5-20/h8-10H,3-7H2,1-2H3,(H,23,24)
SMIL CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CC[N+](C)([O-])CC3
IUPAC navn 1-ethyl-6-fluoro-7-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-4-oxoquinoline-3-carboxylic acid
Molekylvægt (g/mol) 349.36
Mængde 25 mg
Formel vægt 349.1438 g/mol