Få mere at vide
4-(3-aminopropyl)-1,2,5-oxadiazole-3-carboxylic acid 5-oxide, TRC
Artikelnummer. 30316323
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316323 10 mg
30309887 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30316323 Leverandør Toronto Research Chemicals Leverandørnr. TRCA19664010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(3-aminopropyl)-1,2,5-oxadiazole-3-carboxylic acid 5-oxide
CAS 352647-66-6
Molekylær formel C6H9N3O4
Mængde 10 mg
InChI formel InChI=1S/C6H9N3O4/c7-3-1-2-4-5(6(10)11)8-13-9(4)12/h1-3,7H2,(H,10,11)
SMIL NCCCc1c(no[n+]1[O-])C(=O)O
IUPAC navn 4-(3-aminopropyl)-5-oxido-1,2,5-oxadiazol-5-ium-3-carboxylic acid
Molekylvægt (g/mol) 187.15
Formel vægt 187.0593