missing translation for 'onlineSavingsMsg'
Få mere at vide

4-(3,4-Dimethoxyphenyl)butanoic Acid, TRC

Artikelnummer. 30374005
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30374005 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374005 Supplier Toronto Research Chemicals Supplier No. TRCD461090250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-(3,4-Dimethoxyphenyl)butanoic Acid
CAS 13575-74-1
Molekylær formel C12H16O4
Mængde 250 mg
Synonym 3,4-Dimethoxybenzenebutanoic Acid, 4-(3,4-Dimethoxyphenyl)butyric Acid, 3,4-Dimethoxybenzenebutanoic Acid, 4-(3,4-Dimethoxyphenyl)butanoic Acid, 4-(3,4-Dimethoxyphenyl)butyric Acid, NSC 286140
InChI formel InChI=1S/C12H16O4/c1-15-10-7-6-9(8-11(10)16-2)4-3-5-12(13)14/h6-8H,3-5H2,1-2H3,(H,13,14)
SMIL COc1ccc(CCCC(=O)O)cc1OC
Molekylvægt (g/mol) 224.25
Formel vægt 224.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.