Learn More
Memmert™ 4-20 mA Current Loop Interface for UN30
Temperature Output for external temperature monitoring.
Brand: Memmert™ UN30V6
Additional Details : Weight : 0.01200kg
Description
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
UN30 Oven | |
Current Loop Interface | |
4 to 20 mA |