Learn More
Memmert™ 4-20 mA Current Loop Interface for UN160, Actual Value
Temperature Output for external temperature monitoring.
Brand: Memmert™ UN160V3
This item is not returnable.
View return policy
Description
- Temperature controller actual value
Specifications
UN160 Oven | |
Current Loop Interface | |
4 to 20 mA |
Product Content Correction
Product Title
Memmert™ 4-20 mA Current Loop Interface for UN160, Actual Value > For Use With: UN160
Spot an opportunity for improvement?Share a Content Correction