Learn More
Memmert™ 4-20 mA Current Loop Interface for UN160
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UN160V6
Product Code. 17599945
  • 3355.00 DKK / Each
Only null left
Estimated Shipment: 23-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UN160 Oven
Current Loop Interface
4 to 20 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4-20 mA Current Loop Interface for UN160 > For Use With: UN160